Learn More
Invitrogen™ OSBPL11 Monoclonal Antibody (OTI6A6)

Description
OSBPL11 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | OSBPL11 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI6A6 |
| Concentration | 0.78 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | OSBPL11 |
| Gene Accession No. | Q9BXB4 |
| Gene Alias | 9430097N02Rik; AI132306; AU019795; FLJ13012; FLJ13164; ORP11; ORP-11; OSBP12; OSBPL11; OSBP-related protein 11; oxysterol binding protein like 11; oxysterol binding protein-like 11; oxysterol-binding protein-related protein 11; TCCCIA00292; testicular tissue protein Li 132 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.