Learn More
ATCC Oscillospiraceae Bacterium Type 2 Strain KA00274 (ATCC TSD-275), frozen culture
Supplier:  ATCC TSD275
Encompass
Oscillospiraceae bacterium Type 2 (strain KA00274) is a frozen bacterial type strain isolated from human vaginal fluid in 2012. Provided as a frozen culture for research use, this strain grows best in liquid media and typically exhibits little to no turbidity.
  • Type strain isolated from human vaginal fluid in 2012.
  • Frozen culture format suitable for -80°C storage.
  • Preferred growth in liquid media; low turbidity.
  • Documentation available on request: certificate of analysis, safety data sheet, certificate of origin.
  • Intended for research use; handle per institutional biosafety guidelines.
Catalog No. NC3780205