Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0313210
Encompass_Preferred
Oncostatin M (OSM) is a multifunctional cytokine and belongs to Interleukin-6 (IL-6) subfamily which also includes IL-11 leukemia inhibitory factor (LIF) ciliary neurotropic factor cardiotrophin-1 and novel neurotropin-1. In vivo OSM is secreted from activated T cells monocytes neutrophils and endothelial cells. OSM is related to LIF and shares a receptor with LIF in human. Human OSM can bind to gp130 and recruit OSM ReceptorBeta or LIF ReceptorBeta to form a ternary complex. OSM stimulates the growth of different types of cells including megakaryocytes fibroblasts vascular endothelial cells and T cells. OSM inhibits the proliferation of several cancer cell lines such as solid tissue tumor cells lung cancer cells melanoma cells and breast cancer cells.
Catalog No. 50-252-9806
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.