Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Osm-S-106 | 2650846-59-4 | 99.3% | 306.36 g·mol-1 | C12H10N4O2S2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY16223810MG

Encompass_Preferred

OSM-S-106 is a small-molecule pro-inhibitor of Plasmodium falciparum asparagine tRNA synthetase (PfAsnRS). It inhibits protein translation by forming an enzyme-mediated Asn adduct, activates the amino acid starvation response, and shows selective activity against Plasmodium blood and liver stages. Supplied as a high-purity research reagent for parasitology and antimalarial mechanism-of-action studies.

  • High purity (99.34%) suitable for research applications.
  • Well-characterized chemical identity: C12H10N4O2S2, MW 306.36 g·mol-1.
  • Mechanism: reaction-hijacking pro-inhibitor that targets PfAsnRS.
  • Selective activity against Plasmodium blood and liver stages.
  • Provided with product datasheet and analytical information for quality control.
  • Store and handle according to supplier datasheet recommendations.

Catalog No. 50-002-88761


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.