Learn More
Medchemexpress LLC Osm-smi-10b | 2502294-55-3 | 99.7% | 378.33 g·mol⁻¹ | C21H14O7 | 10 MG

Supplier: Medchemexpress LLC HY14869210MG
OSM-SMI-10B is a small-molecule research inhibitor that significantly reduces oncostatin M (OSM)-induced STAT3 phosphorylation in cancer cells. It is intended for biochemical and cellular research into JAK/STAT signaling and STAT3-related pathways. The compound is supplied as a high-purity solid and has defined storage conditions for powder and solutions.
- Significantly reduces OSM-induced STAT3 phosphorylation in cellular assays.
- Directly targets phosphorylated STAT3 (p-STAT3) in the JAK/STAT pathway.
- High purity (99.7%) suitable for research applications.
- Molecular formula C21H14O7; molecular weight 378.33 g·mol⁻¹.
- Powder storage: -20°C for 3 years, 4°C for 2 years.
- In solvent storage: -80°C for 6 months, -20°C for 1 month.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.