Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium ICP Standard, 1000 ppm Os in 20% HCl, Ricca Chemical
SDP

Catalog No. POS1KH500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
POS1KH500 500 mL Natural Poly Bottle
POS1KH100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. POS1KH500 Supplier Ricca Chemical Company Supplier No. POS1KH500
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14996-60-2 , 7647-01-0 , 14996-60-2 , 7732-18-5
Molecular Formula Cl3H2OOs
Molecular Weight (g/mol) 314.60
MDL Number MFCD00149815
InChI Key KHHOLOMEVBPRQY-UHFFFAOYSA-K
PubChem CID 16211485
IUPAC Name trichloroosmium hydrate
SMILES O.Cl[Os](Cl)Cl

Specifications

Packaging Natural Poly Bottle
CAS Min % 79.0, 18.68, 0.15
CAS Max % 82.0, 19.45, 0.15
Physical Form Liquid
Color Black
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg Os (1, 000ppm Os)OsCl3 in 20% HCl
Chemical Name or Material Osmium ICP Standard
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.