Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSMR Monoclonal Antibody (AN-V2), APC, eBioscience™
GREENER_CHOICE

Catalog No. 50245665
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-665 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-245-665 Supplier Invitrogen™ Supplier No. 17130342
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This monoclonal AN-V2 antibody reacts with the human oncostatin M (OSM) receptor. OSM, which belongs to the interleukin-6 cytokine family, is produced by T cells, activated monocytes, and dendritic cells. In humans, OSM can bind two unique receptor complexes. The type I OSM receptor is composed of the LIFR associated with gp130, and recognizes both LIF and OSM. The type II OSM receptor, which consists of gp130 and the OSMR subunit, specifically binds the cytokine, leading to activation of the JAK/STAT signaling pathway. Reports have also indicated a soluble form of the OSM receptor, which this antibody also recognizes. The OSM receptor is expressed in the bone marrow, endothelium, liver, lung, and numerous tumor cell lines. The AN-V2 monoclonal antibody recognizes the type II (beta) OSM receptor. Applications Reported: This AN-V2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AN-V2 antibody has been pre-titrated and tested by flow cytometric analysis of the A375 cell line. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF.Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSMR
Applications Flow Cytometry
Classification Monoclonal
Clone AN-V2
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene OSMR
Gene Accession No. Q99650
Gene Alias IL-31 receptor subunit beta; IL-31R subunit beta; IL-31RB; IL-31R-beta; interleukin-31 receptor subunit beta; MGC150626; MGC150627; MGC75127; oncostatin M receptor; oncostatin receptor; oncostatin-M specific receptor beta subunit; oncostatin-M specific receptor subunit beta; oncostatin-M-specific receptor subunit beta; OSMR; Osmrb; PLCA1
Gene Symbols OSMR
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9180
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.