Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sekisui Diagnostics™ OSOM™ COVID-19 Antigen Control Kit

Catalog No. 22321137 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
COVID-19 Antibodies
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-321-137 COVID-19 Antibodies
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-137 Supplier Sekisui Diagnostics™ Supplier No. 1068

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Meet your external quality control requirements with the OSOM™ COVID-19 Antigen Control Kit.

  • For use with the OSOM™ COVID-19 Antigen Rapid Test
  • Contains 5 Positive Control Swabs and 5 Negative Control Swabs
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes COVID-19 Antibodies
Format 10 Paired Positive/Negative Control Swabs
For Use With (Equipment) Rapid Test
Sample Type Nasal Swab
Type Control Swabs
Disposable Single-use
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.