Learn More
Invitrogen™ OSR1 Monoclonal Antibody (3B6)

Description
Peptide Sequence: MGSKTLPAPV PIHPSLQLTN YSFLQAVNGL PTVPSDHLPN LYGFSALHAV HLHQWTLGYP AMHLPRSSFS KVPGTVSSLV DARFQLPAFP WFPHVIQPKP.
Specifications
Specifications
| Antigen | OSR1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3B6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Osr1 |
| Gene Accession No. | Q8TAX0 |
| Gene Alias | ODD; Odd1; odd-skipped homolog; odd-skipped related 1; odd-skipped related 1 (Drosophila); odd-skipped related transciption factor 1; odd-skipped related transcription factor 1; Osr; OSR1; Protein odd-skipped-related 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.