Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116814 Supplier Invitrogen™ Supplier No. PA5116814
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: NCI-H460, Cal-27, A-549, 22Rv1.

OSR1 is a transcription factor that plays a role in the regulation of embryonic heart and urogenital development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSR1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Osr1
Gene Accession No. B0K011, Q8TAX0, Q9WVG7
Gene Alias ODD; Odd1; odd-skipped homolog; odd-skipped related 1; odd-skipped related 1 (Drosophila); odd-skipped related transciption factor 1; odd-skipped related transcription factor 1; Osr; OSR1; Protein odd-skipped-related 1
Gene Symbols Osr1
Host Species Rabbit
Immunogen Recombinant protein (or fragment)'.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130497, 23967, 298878
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.