Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113150
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113150 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113150 Supplier Invitrogen™ Supplier No. PA5113150
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OSR2 Polyclonal Antibody for ELISA, IHC (P), Western Blot

OSR2 is a mammalian homolog of the Drosophila odd-skipped family of transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSR2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OSR2
Gene Accession No. Q8N2R0
Gene Alias 5430409I15Rik; odd-skipped related 2; odd-skipped related transciption factor 2; Osr2; Osr2A; Osr2B; Protein odd-skipped-related 2
Gene Symbols OSR2
Host Species Rabbit
Immunogen Recombinant Human Protein odd-skipped-related 2 protein (101-276AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116039
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.