Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Osteocalcin Monoclonal Antibody (OC4-30)
GREENER_CHOICE

Catalog No. MA120786 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA120786 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA120786 Supplier Invitrogen™ Supplier No. MA120786
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody does not cross react with mouse Osteocalcin. Predicted molecular weight of 12 kDa.

Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteocalcin
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OC4-30
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and no preservative; pH 7.4
Gene BGLAP
Gene Accession No. P02818, P02822, P04640, P0C225, P39056, P81455, Q8HYY9
Gene Alias Bglap; Bglap1; Bglap2; BGP; Bgpr; Bgpra; bone gamma carboxyglutamate protein; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein; bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; GLA; Gla protein precusor; mOC-A; OC; OCN; OG1; Osteocalcin; PMF1; RP11-54H19.5; unnamed protein product
Gene Symbols BGLAP
Host Species Mouse
Immunogen Bovine osteocalcin.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100340503, 100861188, 25295, 396348, 397530, 403762, 632, 677670
Target Species Human, Rat, Canine, Rabbit, Sheep, Pig, Chicken, Goat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.