Learn More
Cellular Engineering Technologies Osteogenic Differentiation Media (450mL)
Supplier:  Cellular Engineering Technologies MR1009
Encompass_Preferred
Human Osteogenic Differentiation Media is designed to support the differentiation of adipose or bone marrow-derived mesenchymal stem cells into osteogenic or bone-producing cells. Media is compatible with available animal-origin serum, supplemented separately according to the customer preference.Media is shipped with gel packs.
Catalog No. NC2753078