Learn More
IBL America Osteopontin N-Half (Mouse) ELISA
Supplier:  IBL America 27259
Encompass_Preferred
Solid phase sandwich ELISA for the determination of mouse OPN N-Half in EDTA-plasma, urine or cell culture media. For research use only, not for use in diagnostic procedures.As a primary antibody is coated on a plate, samples and standard are added into the wells for the 1st reaction. After the reaction, HRP-conjugated secondary antibody is added into the wells for 2nd reaction. After washing away unbound the secondary antibody, Tetra Methyl Benzidine (TMB) is added to the wells and color develops.
Catalog No. 50-136-6512
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.