Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Osteopontin/OPN Antibody Blocking Peptide
SDP

Catalog No. NB6001043E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001043E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001043E Supplier Novus Biologicals™ Supplier No. NB6001043PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide from human Osteopontin/OPN. Source: Synthetic The Osteopontin/OPN Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 6696
Species Human
Protein Family Osteopontin/OPN
Purity >95%
Accession Number P10451
Concentration 1.0 mg/mL
Content And Storage Store at -20 C short term. Aliquot and store (at -80 C long term. Avoid freeze-thaw cycles.)
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
For Use With (Application) Antibody Competition
Gene Symbol SPP1
Quantity 0.05 mg
Regulatory Status RUO - research use only
Research Category Cellular Markers
Source Synthetic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.