Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSTN Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734787 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4787 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4787 Supplier Proteintech Group Inc Supplier No. 252071AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen OSTN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene OSTN
Gene Accession No. P61366
Gene Alias MUSCLIN, osteocrin, OSTN
Gene Symbols OSTN
Host Species Rabbit
Immunogen OSTN Fusion Protein Ag19166
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344901
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.