Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OTP Monoclonal Antibody (CL11225)
GREENER_CHOICE

Catalog No. PIMA544172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544172 Supplier Invitrogen™ Supplier No. MA544172
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The antibody solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VKCRLGVGGS DPGGHPGDLA PNSDPVEGAT LLPGEDITTV GSTPASLAVS AKDPDKQPGP QGGPNPSQAG QQQGQQKQKR HRTRFTPAQL NELE Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

This gene encodes a member of the homeodomain (HD) family. HD family proteins are helix-turn-helix transcription factors that play key roles in the specification of cell fates. This protein may function during brain development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone CL11225
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OTP
Gene Accession No. Q5XKR4
Gene Alias homeobox protein orthopedia; orthopedia homeobox; orthopedia homolog; orthopedia homolog (Drosophila); Otp
Gene Symbols OTP
Host Species Mouse
Immunogen Recombinant protein corresponding to Human OTP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23440, 294640
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.