Learn More
Selleck Chemical LLC OTUB2-IN-1-E1880-25MG
SDP
Supplier:  Selleck Chemical LLC E188025MG
Encompass_Preferred
OTUB2-IN-1 is a specific inhibitor of OTUB2 decreases PD-L1 protein expression in tumor cells leading to suppressed tumor growth through enhanced infiltration of cytotoxic T lymphocytes (CTLs) within the tumor microenvironment
Catalog No. 50-313-7711
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.