Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OTX 015
GREENER_CHOICE

Catalog No. 618110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-811-0 10 mg
61-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-811-0 Supplier Tocris Bioscience™ Supplier No. 6181/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

BET bromodomain inhibitor; orally bioavailable

BET bromodomain inhibitor (IC50 values are in the range 92 - 112 nM). Inhibits proliferation and induces apoptosis of leukemia cell lines in vitro. Decreases BRD2, BRD4 and c-MYC protein expression. Orally bioavailable.

Chemical Identifiers

CAS 202590-98-5
Molecular Formula C25H22ClN5O2S

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight 491.99
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 491.99
Chemical Name or Material (6S)-4-(4-Chlorophenyl)-N-(4-hydroxyphenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.