Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite Outdoor Flammable Cabinet
GREENER_CHOICE

Catalog No. 19033905 Shop All Justrite Safety Group Products
Change view
Click to view available options
Dimensions (D x W x H):
18 x 43 x 44 in.
18 x 43 x 65 in.
Door Style:
Manual-close, Hinged, Three-point lock, Locking
Self-close, Hinged, Three-point lock, Locking, Auto-close
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Door Style
19-033-905 18 x 43 x 65 in. Self-close, Hinged, Three-point lock, Locking, Auto-close
19-033-906 18 x 43 x 65 in. Manual-close, Hinged, Three-point lock, Locking
19-033-907 18 x 43 x 44 in. Self-close, Hinged, Three-point lock, Locking, Auto-close
19-033-908 18 x 43 x 44 in. Manual-close, Hinged, Three-point lock, Locking
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-033-905 Supplier Justrite Supplier No. 814520
Add to Cart
Add to Cart

Engineered to withstand the elements and potential impacts, this cabinet is OSHA, NFPA, and FM approved, ensuring your flammable chemicals are safely and securely stored outdoors.

  • Spill Slope shelves with fully welded sump
  • Ground Screw
  • Durable and Chemical - resistant powder coated paint
  • UV resistant
  • Vent with built in flash arrestors
  • Welded Shelf Hangers
  • Leak Proof Sump

Specifications

Certifications/Compliance OSHA, NFPA, and FM approved
Description 45 gal.
Shelf Type Adjustable
Color Yellow
Material Steel (18 ga)
For Use With (Application) Safe outdoor storage of flammable liquids to avoid risk of potential fire hazards and chemical spills in industries like chemical manufacturing, petrochemical and laboratories
Product Type Flammables Storage Cabinet
Door Style Self-close, Hinged, Three-point lock, Locking, Auto-close
Dimensions (D x W x H) Interior 14.6 x 39.5 x 60 in.
Dimensions (D x W x H) 18 x 43 x 65 in.
Weight (English) 264.7 lb.
No. of Doors 2
No. of Shelves 2
Weight (Metric) 120 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.