Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Outlander™ Safety Eyewear

Sporty dual frame style with wide temples, suitable for both working environments and sports activities

Specifications

Certifications/Compliance ANSI Z87.1
Frame Color Black
Weight (Metric) 32 g
Lens Material Polycarbonate
Frame Material PC and PET
View More Specs

Products 4
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-155-827
View Documents
Pyramex™
SB8010D
Clear N/A N/A N/A
19-155-828
View Documents
Pyramex™
SB8020D
Gray N/A N/A N/A
19-155-829
View Documents
Pyramex™
SB8065D
Blue, Mirror N/A N/A N/A
19-155-830
View Documents
Pyramex™
SB8070D
Silver, Mirror N/A N/A N/A
Description

Description

  • Sporty dual lens style with wide temples
  • Soft nose piece provides comfort for extended use without slipping
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications

    Specifications

    ANSI Z87.1
    32 g
    PC and PET
    Black
    Polycarbonate
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.