Learn More
Carbosynth LLC. OVA Peptide (323-339), 5mg, An immune checkpoint peptide, MW: (M.W. 1773.94), Molecular formula: C74H120N26O25, CAS# [92915-79-2]
Supplier: Carbosynth LLC. POV3636PI5MG
Synonyms: H-Ile-Ser-Gln-Ala-Val-His-Ala-Ala-His-Ala-Glu-Ile-Asn-Glu-Ala-Gly-Arg-OH; Chicken ovalbumin fragment (323-339), ISQAVHAAHAEINEAGR, Application: T-cell Activating Peptide, Storage Temperature: -20°C, References: F. De Mattia, S. Chomez, F. Van Laethem, V. Moulin, J. Urbain, M. Moser, O. Leo, and F. Andris, J. Immunol., 163, 5929, (1999), S.-J. Sung, C.E. Rose, and S.M. Fu, J. Immunol., 166, 1261, (2001).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.