Learn More
Worthington Biochemical Corporation OVALBUMIN 5GM
SDP
Supplier:  Worthington Biochemical Corporation LS003048
Encompass_Preferred
Major protein of egg white, with a molecular weight of 45 kDa. A lyophilized powder. 9006-59-1
Catalog No. 50-592-371
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.