Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ovalbumin Native Protein
SDP

Catalog No. P5241 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P5241 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P5241 Supplier R&D Systems™ Supplier No. P5241

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Native Protein

Specifications

Concentration LYOPH
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 396058
Molecular Weight (g/mol) 45 kDa
Quantity 1 g
Storage Requirements Store at -4°C in powder form. Store at -20°C once reconstituted.
Gene Alias Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8
Gene Symbol OVAL
Reconstitution Reconstitute in sterile water at 1-10mg/ml. We do not recommend reconstituting entire 1g, instead you may weight out a smaller amount, reconstitute it, aliquot, and store at -20/-80°C. The remainign powder should remain stored at 4°C.
Inhibitor Properties Ovalubin is very stable in the lyophilized state, we recommend making (1-10mg/ml) solutions as needed. Frozen stocks of ovalbumin in water at 5 mg/ml have been stable in QC for at least a month. Reconstituted stability is diluent, concentration, technique, and application specific.
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Ovalbumin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.