Learn More
Hooke Laboratories Inc Ovalbumin (OVA) in PBS, 2mg in 1mL
Supplier:  Hooke Laboratories Inc DS0142
Encompass
Chicken ovalbumin (OVA) protein. 2 mg in 1 mL of phosphate buffered saline (PBS). Sterile filtered. Supplied in vials, ready to use
Catalog No. NC1673368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.