Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ovarian Carcinoma Antigen Monoclonal Antibody (OV632)
GREENER_CHOICE

Catalog No. 01672046
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-672-046 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-046 Supplier Invitrogen™ Supplier No. MA135740
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody stains 80% of non-mucinous primary and metastatic ovarian cancers. This monoclonal antibody is used for the identification of primary and metastatic non-mucinous ovarian carcinoma and ovarian carcinoma cells in peritoneal fluids. It rarely stains nongynaecological malignancies.

OA3 is an ovarian carcinoma membrane antigen which is present in ovarian carcinomas and in cyst fluids obtained from these carcinomas, as well as in oviductal carcinomas and some endometrial carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ovarian Carcinoma Antigen
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OV632
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias gynecologic tumor; ovarian carcinoma; ovary cancer
Host Species Mouse
Immunogen Cyst fluid of a human serous cystadenocarcinoma of the ovary.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.