Learn More
Description
Specifications
Specifications
| Formulation | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Quantity | 5 Slides |
| Storage Requirements | −20°C short term. Aliquot and store at −80°C long term. Avoid freeze-thaw cycles |
| Product Type | Human Ovary Tissue Lysate (Adult) |
| Cross Reactivity | Human |
| Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
