Learn More
VistaLab Technologies™ Ovation™ Adjustable-Volume Pipetters PROMO
SDP
Naturally follows the contours of the palm.
Supplier:  VistaLab Technologies™ 10570002TI
Catalog No. 22-146-600TI
Only null left
Add to Cart
Description
Description
  • The adjustable hook can be easily positioned for a custom fit for left- or right-handed users
  • Fits comfortably in the palm of your hand
  • Volume setting is indicated by an LCD readout on the pipetter handle
  • Significantly improves postures and reduces forces associated with pipetting
  • Five popular volume settings can be programmed and recalled by pressing a single button
  • Energy release button stores the energy captured during the acquisition of tips and uses energy to push off the tip at the gentle push of a button
  • Low-force, form-fitting plunger allows user to manually control the speed of aspirating and dispensing with minimal effort
  • Nozzle with replaceable filter prevents contamination of the pipetter
  • Stands securely on the counter before, during, and after the job—no risk of contamination
  • In-lab calibration adjusted with internal software
Product Suggestions
Product Suggestions