Learn More
Restek™ Oven Flapper Assemblies
SDP
Catalog No. 06611246 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890 GCs
Agilent 6890 GCs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
06-611-246 Agilent 6890 GCs
06-611-245 Agilent 5890 GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-246 Supplier Restek™ Supplier No. 22995
Ensure even and consistent column heating when the insulation becomes damaged or worn, for Agilent GCs
  • Replacement Insulation Gasket and Hardware Kit includes Insulation Gasket (1), Screws (2) and Nuts (2)
  • Insulation is replaceable, using Restek's Oven Flapper Assemblies
  • Meets or exceeds original manufacturer's performance
Specifications
Product Type Oven Flapper Assembly
For Use With (Equipment) Agilent 6890 GCs