Learn More
PIP™ OverSite™ OTG Rimless Safety Glasses with Black/Gray Temple, Clear Lens
Catalog No. 19161646 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Disposable:
Reusable
Catalog No. Disposable
19-161-646 Reusable
1 options
This item is not returnable. View return policy
Catalog No. 19-161-646 Supplier PIP™ Supplier No. 250980000
  • $18.00
    $12.65 / Pair
    Save $5.35
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,249.00
    $1,585.65 / Case of 144 PR
    Save $663.35
    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adjustable Over-The-Glass (OTG) style offers a perfect fit over any kind of personal eyewear.
  • Clear lens
  • Over-the-glass design helps keep prescription eyewear users in compliance
  • Can also be worn by itself as a regular safety glass
  • Rimless frame design ensures an unobstructed view
  • Adjustable length temples are rubber co-molded, offering the user a secure custom fit
  • Ratchet temples for better angle adjustment
  • Molded nose bridge design to fit the vast majority of users
  • U-Fit spatula temples
  • Meets ANSI Z87.1+
  • Meets CSA Z94.3
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Over-the-Glasses Safety Glasses
For Use With (Application) Fit over any kind of personal eyewear
Frame Color Black/Gray
Temple Style U-Fit™ spatula temples
Weight (Metric) 39 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 99.99% UVA and UVB light absorption
Frame Type Rimless