Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OVGP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520580
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520580 Supplier Invitrogen™ Supplier No. PA520580
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Despite its predicted molecular weight, OVGP1 often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human placenta lysate. PA5-20580 can be used with blocking peptide PEP-0700.

Oviductins belong to a family of glycoproteins that have been suggested to play several roles during the early processes of reproduction. OVGP1 is a large carbohydrate-rich, epithelial glycoprotein with numerous O-glycosylation sites within threonine, serine, and proline-rich tandem repeats. The protein is secreted from non-ciliated oviductal epithelial cells and associates with ovulated oocytes, blastomeres, and spermatozoan acrosomal regions. At least two isoforms of OVGP are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OVGP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene OVGP1
Gene Accession No. Q12889, Q62010
Gene Alias 120kDa; AU016433; AU019448; CHIT5; chitinase 5; EGP; Estrogen-dependent oviduct protein; MOGP; MUC9; mucin 9; mucin-9; OGP; Ovgp1; oviduct glycoprotein; oviduct glycoprotein OGP; Oviductal glycoprotein; oviductal glycoprotein 1; oviductal glycoprotein 1, 120kD; oviductal glycoprotein 1, 120kDa; oviductin; oviduct-specific glycoprotein
Gene Symbols OVGP1
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the amino terminus of human OVGP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12659, 5016
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.