Learn More
Kingfisher Biotech Inc Multi-species IL-12 p40 (Caprine, Ovine)Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1151V025
Encompass_Preferred
The Ovine/Caprine IL-12/IL-23 p40 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ovine/Caprine IL-12/IL-23 p40 applications are for cell culture, ELISA standard, and Western Blot Control. Ovine/Caprine IL-12/IL-23 p40 yeast-derived recombinant protein can be purchased in multiple sizes. Ovine/Caprine IL-12/IL-23 p40 Specifications: (Molecular Weight: 34.5 kDa) (Amino Acid Sequence: IWELEKNVYV VELDWYPNAP GETVVLTCDT PEEDGITWTS DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSRSLL LLHKKEDGIW STDILKDQKE PKAKSFLKCE AKDYSGHFTC SWLTAISTNL KFSVKSSRGS SDPRGVTCGA ASLSAEKVSM DHREYNKYTV ECQEGSACPA AEESLPIEVV MEAVHKLKYE NYTSSFFIRD IIKPDPPKNL QLRPLKNSRQ VEVSWEYPDT WSTPHSYFSL TFCVQVQGKN KREKKLFTDQ TSAKVTCHKD ANIRVQARDR YYSSFWSEWA SVSCS (305)) (Gene ID: 443472). For research use only. Made in the USA
Catalog No. 50-253-3613
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.