Learn More
Kingfisher Biotech Inc Ovine CCL3 Recombinant Protein
Supplier: Kingfisher Biotech Inc RP1529V005
The Ovine CCL3 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ovine CCL3 applications are for cell culture, ELISA standard, and Western Blot Control. Ovine CCL3 yeast-derived recombinant protein can be purchased in multiple sizes. Ovine CCL3 Specifications: (Molecular Weight: 7.8 kDa) (Amino Acid Sequence: APFGADTPTA CCFSYAARQL SRKIVADYFE TSSQCSKPGV IFQTKRGRQV CANPSEDWVQ QYVTDLELNS (70)) (Gene ID: 101114535). For research use only. Made in the USA
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.