Learn More
Kingfisher Biotech Inc Ovine CD40 Ligand (CD40LG) Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1638V500
Encompass_Preferred
The Ovine CD40 Ligand yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ovine CD40 Ligand applications are for cell culture, ELISA standard, and Western Blot Control. Ovine CD40 Ligand yeast-derived recombinant protein can be purchased in multiple sizes. Ovine CD40 Ligand Specifications: (Molecular Weight: 16.1 kDa) (Amino Acid Sequence: MHKGDQEPQI AAHVISEASS KTTSVLQWAP KGYYTLSSNL VTLENGKQLA VKRQGFYYIY TQVTFCSNRE ALNQAPFIAS LCLKSPSGSE RILLRAANTH SSSKPCGQQS IHLGGVFELQ QGASVFVNVT DPSQVSHGTG FTSFGLLKL (149)) (Gene ID: 767628). For research use only. Made in the USA
Catalog No. 50-253-3016
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.