Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ovine IP-10 ELISA Kit

Catalog No. ESH6RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ESH6RB 96 Tests
ESH6RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ESH6RB Supplier Invitrogen™ Supplier No. ESH6RB
Add to Cart
Add to Cart

ELISA

Ovine IP-10 quantitates ovine IP-10 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant ovine IP-10.

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C x C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; crg-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamma induced protein) (Gamma-IP10) (IP-10); small inducible cytokine B subfamily (Cys-X-Cys) member 10; small inducible cytokine B subfamily (Cys-X-Cys), member 10; small inducible cytokine subfamily B (Cys-X-Cys), member 10; small-inducible cytokine B10
Assay Range 28 to 7,000 pg/mL
Assay Sensitivity 28 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 442997
Gene Symbol CXCL10
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CXCL10 (IP-10)
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.