Learn More
Kingfisher Biotech Inc Ovine LIF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2132V100
Encompass_Preferred
The Ovine LIF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Ovine LIF applications are for cell culture, ELISA standard, and Western Blot Control. Ovine LIF yeast-derived recombinant protein can be purchased in multiple sizes. Ovine LIF Specifications: (Molecular Weight: 19.8 kDa) (Amino Acid Sequence: SPLPITPVNA TCATRHPCPS NLMNQIRNQL GQLNSSANSL FILYYTAQGE PFPNNLDKLC SPNVTDFPPF HANGTEKARL VELYRIIAYL GASLGNITRD QKVLNPYAHS LHSKLNATAD VLRGLLSNVL CRLCSKYHVS HVDVTYGPDT SGKDIFQKKK LGCQLLGKYK QVIAVLAQAF (180)) (Gene ID: 101110180). For research use only. Made in the USA
Catalog No. 50-253-2382
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.