Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ A2 Large Gel Systems with Buffer Exchange Ports PROMO

Catalog No. 09528102PM Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions (L x W x H):
14.8 x 12.8 x 4.1 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
09-528-102PM 14.8 x 12.8 x 4.1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-528-102PM Supplier Thermo Scientific™ Supplier No. A2BPPM

missing translation for 'validMainframeMsg'

Handle high throughput with consistent, reliable results with Thermo Scientific™ Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.

Features:

  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (16-, 24- and 36-well), 1.5 mm thick
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Complete system with buffer exchange ports
Product Line Owl
Sample Size 8 to 432 (16-, 24- and 36-well combs)
Volume (Metric) Buffer 2.3 L
Dimensions (L x W x H) 14.8 x 12.8 x 4.1 in.
Length (English) 14.8 in.
Length (Metric) 37.5 cm
Length (Metric) Gel 25 cm
Width (English) 12.8 in.
Width (Metric) 32.5 cm
Width (Metric) Gel 20 cm
Height (English) 4.1 in.
Height (Metric) 10.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.