Learn More
Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts
Catalog No. 09528204
Change view
Click to view available options
Type:
End Gates
Gasket
Gel Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-528-204 End Gates
OW-A3-GK-1 Gasket
OW-A3UVT1 Gel Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-528-204 Supplier Thermo Scientific™ Supplier No. A31EG
Add to Cart
Add to Cart
Choose these replacement parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
Choose Thermo Scientific™ Owl™ A3-1 and A6 Wide Gel System Replacement Parts, which include a gasketed UVT gel tray, replacement gaskets and end gates.
The gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run utilizing a wall comb. Built in buffer exchange ports allows buffer recirculation on extended runs, if needed. The large number of comb slots gives you sample and resolving distance flexibility.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ A3-1 and A6 Wide Gel System
Product Line Owl
Type End Gates
Quantity 1 Pair