Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ ADJ3 Adjustable System Accessories

Catalog No. OWADJ365LM
Change view
Click to view available options
Type:
Gasket
Heat Sink
Size:
65 x 35 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Size
OW-ADJ365LM Heat Sink 65 x 35 cm
OW-ADJ3GK Gasket -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OW-ADJ365LM Supplier Thermo Scientific™ Supplier No. ADJ365LM
Add to Cart
Add to Cart

Owl ADJ3 Adjustable System is the widest adjustable system perfect for sequencing and proteins.

  • The height of the upper buffer chamber (UBC) may be adjusted from 20 cm to 80 cm by sliding the UBC up or down two support poles. The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) ADJ1 Giraffe™ Adjustable Gel Electrophoresis System
Type Heat Sink
Size 65 x 35 cm
Thickness (English) 0.18 in.
Thickness (Metric) 0.48 cm
Dimensions (L x W) 25.5 x 13.7 in. (65 x 35 cm)
Length (English) 25.5 in.
Length (Metric) 65 cm
Width (English) 13.7 in.
Width (Metric) 35 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.