Learn More
Thermo Scientific™ Owl™ ADJ3 Adjustable System Accessories
Catalog No. OWADJ3GK Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Gasket
Heat Sink
Size:
65 x 35 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Size
OW-ADJ3GK Gasket -
OW-ADJ365LM Heat Sink 65 x 35 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OW-ADJ3GK Supplier Thermo Scientific™ Supplier No. ADJ3GK
Owl ADJ3 Adjustable System is the widest adjustable system perfect for sequencing and proteins.
  • The height of the upper buffer chamber (UBC) may be adjusted from 20 cm to 80 cm by sliding the UBC up or down two support poles. The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ADJ1 Giraffe™ Adjustable Gel Electrophoresis System
Type Gasket