Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ Blotting Paper

Catalog No. p-4525105
Change view
Click to view available options
Length (Metric):
9 cm
10 cm
12 cm
18 cm
20 cm
35 cm
46 cm
Width (Metric):
9 cm
10 cm
16 cm
20 cm
45 cm
57 cm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Width (Metric)
OW-FP-1 20 cm 20 cm
OW-FP-2 35 cm 45 cm
OW-FP-3 46 cm 57 cm
OW-FP-4 10 cm 10 cm
OW-FP-6 9 cm 9 cm
OW-FP-7 12 cm 16 cm
OW-FP-5 18 cm 20 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. OW-FP-1 Supplier Thermo Scientific™ Supplier No. FP1
Only null left
Add to Cart
Add to Cart

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.

  • High purity, 100% cotton fiber
  • Available in pre-cut mini gel and large format gel sizes
  • Compatible with Owl electroblotting systems

Ordering Information:

  • Sold in packs of 100 sheets
TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 20 cm
Width (Metric) 20 cm
Packaging Type Sheets
Flow Rate 35 mL/min.
For Use With (Equipment) Owl electroblotting systems
Weight 183 g/m2
Format Sheet
Material 100% Cotton Fiber
Dimensions (LxW) 20 cm x 20 cm
Thickness 0.83 mm
Product Line Owl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.