Learn More
Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs
Catalog No. OWC27D Shop All Thermo Scientific Products
Change view
Click to view available options
Teeth Count:
7
Thickness (Metric) Teeth:
1.0 mm
1.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Teeth Count Thickness (Metric) Teeth
OW-C27D 7 1.5 mm
OW-C27C 7 1.0 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OW-C27D Supplier Thermo Scientific™ Supplier No. C27D
Eliminate comb assembly with these heavy-duty, one-piece, standard seven-tooth combs.
Eliminate comb assembly with heavy-duty, one-piece Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Combs. These standard seven-tooth combs are available in two thicknesses (1.0 and 1.5mm) and are for use with the Owl™ C2-S Micro Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.
TRUSTED_SUSTAINABILITY
Specifications
Description Standard comb
For Use With (Equipment) Owl™ C2-S Micro Electrophoresis System
Product Line Owl
Type Comb
Teeth Count 7
Thickness (Metric) Teeth 1.5 mm