Learn More
Description
Screen up to seven samples on one gel in less than five minutes with the Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System.
- Horizontal microgel system is designed for quick restriction fragment analysis.
- Microgel may also be used to quickly check a few PCR samples before loading a large number of samples on a larger gel, saving time
- Glass slides, 2 × 3 in. (5 × 7.5cm), serve as the gel tray
- To use, pour and form thin gels by surface tension directly on glass slide; complete run lasts five to 10 minutes
- Using available caster and casting slides, gels can be cast and solidified in less than 10 minutes.
- Buffer chamber (buffer volume, 100mL)
- Lid
- Power supply leads
- Casting tray
- 10 casting slides
- One seven-well comb, 1.5mm thick
Specifications
Specifications
| Description | Horizontal Electrophoresis System; Gel size: 5 x 7.5cm |
| Gel Size | 5 x 7.5 cm |
| Product Line | Owl |
| Type | Micro |
| Sample Size | 7 |
| Volume (Metric) Buffer | 100 mL |
| Dimensions (L x W x H) | 6.75 x 2.63 x 4.38 in. (17 x 38 x 11 cm) |
| Length (English) | 6.75 in. |
| Length (Metric) | 17 cm |
| Length (Metric) Gel | 7.5 cm |
| Show More |
Frequently Asked Questions (FAQs)
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.