Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts
Catalog No. OWD2CST
Change view
Click to view available options
Type:
Caster
Gasket
Gel Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
OW-D2CST Caster
OW-D2GK Gasket
09-528-166 Gel Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. OW-D2CST Supplier Thermo Scientific™ Supplier No. D2CST
Add to Cart
Add to Cart
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
TRUSTED_SUSTAINABILITY
Specifications
Description External caster (no trays/combs)
For Use With (Equipment) Owl™ D2 Wide Gel Electrophoresis System
Product Line Owl
Type Caster