Description
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
Specifications
Specifications
| For Use With (Equipment) | Owl™ D2 Wide Gel Electrophoresis System |
| Product Line | Owl |
| Type | Gasket |
| Quantity | 1 Pair |