Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ D3-14 Wide Gel Electrophoresis System
GSA_VA

Catalog No. 09528128 Shop All Thermo Scientific Products
Change view
Click to view available options
Gel Size:
14 x 23 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Size
09-528-128 14 x 23 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-528-128 Supplier Thermo Scientific™ Supplier No. D314
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Screen PCR products, plasmid preps, restriction mapping and cloning with the Thermo Scientific™ Owl™ D3-14 Wide Gel Electrophoresis System.

Screen PCR products, plasmid preps, restriction mapping and cloning with the Thermo Scientific™ Owl™ D3-14 Wide Gel Electrophoresis System.

Features:
  • Screen 25 to 200 samples on a single agarose gel in less than 30 minutes, producing clear, tight banding patterns with no “smiling.”
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96, eight or 12-well plate.
  • Incorporates Owl™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
  • Sample loading with multichannel pipetter speeds up transfer.
  • Electrode placement allows gel to be run quickly without compromising resolution.
  • Bubble level ensures casting of flat, uniformly thick gels.

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power cords
  • Thermo Scientific™ EasyCast™ gasketed UVT gel tray
  • External caster
  • Four combs (50 micro well, 1.5mm thick)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Gel size L x W: 14 x 23cm; Buffer volume: 800mL; L x W x H: 30 x 27 x 11cm
Gel Size 14 x 23 cm
Product Line Owl
Type Wide
Sample Size 25 to 200
Volume (Metric) Buffer 800 mL
Dimensions (L x W x H) 11.8 x 10.6 x 4.3 in. (30 x 27 x 11 cm)
Length (English) 11.8 in.
Length (Metric) 30 cm
Length (Metric) Gel 14 cm
Width (English) 10.6 in.
Width (English) Gel 6.5
Width (Metric) 27 cm
Width (Metric) Gel 23 cm
Height (English) 4.3 in.
Height (Metric) 11 cm
Show More Show Less
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.