Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis System with Buffer Exchange Ports PROMO

Catalog No. 09528118PM Shop All Thermo Scientific Products
Change view
Click to view available options
Gel Size:
Mini
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Size
09-528-118PM Mini
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-528-118PM Supplier Thermo Scientific™ Supplier No. B2BPPM

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve superior flexibility in a small footprint with our B2 mini gel electrophoresis systems.

  • Two comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands

Ordering Information:

Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed U.V. Transmissible (UVT) gel tray
  • Two combs: 12 and 20 well, double-sided, 1.0/1.5 mm thick
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description EasyCast w/buffer exchange ports
Gel Size Mini
Product Line Owl
Type Mini Gel Electrophoresis System
Sample Size 8 to 48
Volume (Metric) Buffer 800 mL
Dimensions (L x W x H) 9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5 cm)
Length (English) 9.63 in.
Length (Metric) 24.5 cm
Length (Metric) Gel 14 cm
Width (English) 7.06 in.
Width (Metric) 18 cm
Width (Metric) Gel 12 cm
Height (English) 3.75 in.
Height (Metric) 9.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.