Learn More
Thermo Scientific™ Owl™ Leveling Platform
Catalog No. 09528177 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Bubble Level
Leveling Platform
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-528-177 Leveling Platform
09-528-162 Bubble Level
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-528-177 Supplier Thermo Scientific™ Supplier No. BLP
Accommodate most applications with this three-point leveling platform which ensures a flat casting and running surface.
Accommodate most applications with the Thermo Scientific™ Owl™ Leveling Platform, a three-point leveling platform that ensures a flat casting and running surface.
  • Size: 36 x 46cm
  • Includes bubble level
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl System
Product Line Owl
Type Leveling Platform