Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ P9DS Dual Gel System Side cCamps

Catalog No. FBVE1610 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Gasket
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
FB-VE16-10 Gasket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB-VE16-10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Replace blocking plate, buffer chambers, lid or side clamps on your dual gel system with Fisherbrand™ P9DS Dual Gel System Replacement Parts.

Replace blocking plate, buffer chambers, lid or side clamps with Fisherbrand™ P9DS Dual Gel System Replacement Parts, for use with the P9DS Dual Gel System.

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
For Use With (Equipment) Fisherbrand P9DS Gel Electrophoresis System
Type Gasket
Quantity 1 Pair
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.