Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix Owl Pellet
SCI_ED

Catalog No. S00216 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Length (English):
1.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S00216 1.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00216 Supplier AquaPhoenix Supplier No. OW5788P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterilized owl pellet individually wrapped in foil.

  • Approximately 1.5 to 2 in.
  • Individually wrapped in foil
  • Sterilized

Specifications

For Use With (Application) Educational Use
Includes Single Pellet
Length (English) 1.5 in.
Length (Metric) 3.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.